Putket, koeputket ilman reunusta, paksuseinäinen
Toimittaja: SciLabware
1626/20M S/3L6016 1626/10M 1626/12M
212-7431EA 525 EUR
212-7431 212-7454 212-7428 212-7429
Putket, koeputket ilman reunusta, paksuseinäinen
Putket Koeputket
Pyrex®, borosilicate glass, heavy wall.
  • Excellent chemical resistance
  • High temperature resistance
High quality test tubes designed and constructed for optimum performance. Rimless test tubes with a heavy wall to provide extra mechanical strength. Improved user safety.
Sertifikaatit: ISO 4142