VWR®, Stoppers, PE
Toimittaja: VWR Collection
212-0550EA 58.1 EUR
212-0550 212-0551 212-0552 212-0553 212-0554 212-0572 212-0573 212-0574 212-0575 212-0576 212-0577 212-0578 212-0664 212-0665 212-0666 212-0667 212-0679
VWR®, Stoppers, PE
Tulpat PE/LDPE/HDPE
Stopper for test tubes Ø 12 to 16 mm, polyethylene, with tongues.
  • Choice of colours