Anti-Alpaca IgG Goat Polyclonal Antibody (CF®680)
Toimittaja: Biotium
20887-100UL
20887-100ULEA 275 EUR
20887-100UL
Anti-Alpaca IgG Goat Polyclonal Antibody (CF®680)
Vasta-aineet
Goat anti-alpaca IgG antibody that recognizes the VHH domain of heavy chain alpaca IgG. Labeled with CF® Dyes and suitable for western or IF applications.
  • Available in 6 bright and photostable CF® Dyes
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues
  • CF® Dye and biotin conjugates: add 0.5 mL distilled water
  • HRP or DNP conjugates: add 1 ml distilled water
Goat Anti-Alpaca IgG, VHH antibody is an affinity-purified polyclonal goat IgG whole antibody that recognizes the VHH domain of heavy chain alpaca IgG, subclasses 2 and 3,More Product Information