GC septa, Thermogreen® LB-1
Toimittaja: Merck
20657-U 20658 20661
20657-UEA 155 EUR
20657-U SUPL20658 20661.
GC septa, Thermogreen® LB-1
Septumi
These septa are suitable for use with 50 to 300 °C inlet temperatures.