Fosforihappo ≥85% Ph. Eur.

Toimittaja: VWR Chemicals
Vaara

20626.292EA 140 EUR
20626.292 20626.361 20626.463
Fosforihappo ≥85% Ph. Eur.
Fosforihappo
Kaava: H₃PO₄
Sulamispiste: 28 °C
Tiheys: 1,689 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011340
CAS-numero: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III

Order Now

Testitulosten erittely

Assay 84.0 - 90.0 %
Appearance Conforms (see CoA/CoS)
Identification A Passes test
Identification B Passes test
Appearance of solution Passes test
Substances precipitated with ammonia Passes test
Hypophosphorous and phosphorous acids Passes test
Cl (Chloride) Max. 50 ppm
SO4 (Sulphate) Max. 100 ppm
As (Arsenic) Max. 2 ppm
Fe (Iron) Max. 50 ppm
Heavy metals (as Pb) Max. 10 ppm
Residual solvents Unlikely by manuf.process


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR