Anti-IgG Llama Polyclonal Antibody (CF™ 647)

Toimittaja: Biotium
20458-50UL 20458-500UL
20458-50ULEA 127 EUR
20458-50UL 20458-500UL
Anti-IgG Llama Polyclonal Antibody (CF™ 647)
Vasta-aineet
Llama anti-mouse CF™ dye conjugates are prepared by labeling affinity purified llama anti-mouse IgG (H+L) with a selection of CF™ dyes. CF™ dyes offer combined advantages in brightness, photostability, and specificity compared other fluorescent dyes. Far-red fluorescent CF™647 (Ex/Em 650/665 nm) is brighter than Cy®5 and an excellent alternative to Alexa Fluor® 647 for antibody conjugates.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF647
Epitope:
Host: Llama
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR