Anti-IgG Goat Polyclonal Antibody (CF™ 640R)

Toimittaja: Biotium
20304 20304-1
20304.EA 466 EUR
20304. 20304-1
Anti-IgG Goat Polyclonal Antibody (CF™ 640R)
Vasta-aineet
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 640R Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR