Anti-IgG Goat Polyclonal Antibody (CF™ 770)

Toimittaja: Biotium
20288 20288-1
20288.EA 359 EUR
20288. 20288-1
Anti-IgG Goat Polyclonal Antibody (CF™ 770)
Vasta-aineet
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF® 770 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR