Sitruunahappo, anhydrous 99.5-100.5%, powder Ph. Eur.

Toimittaja: VWR Chemicals
Varoitus

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

20282.293EA 95.1 EUR
20282.293 20282.362 20282.464
Sitruunahappo, anhydrous 99.5-100.5%, powder Ph. Eur.
Sitruunahappo

Varoitus: Not suitable for parenteral use

Kaava: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Kiehumispiste: 310
Sulamispiste: 149…151 °C
Tiheys: 1.665 g/cm³ (18 °C. OECD 109)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011669
CAS-numero: 77-92-9
EINECS: 201-069-1
REACH: 01-2119457026-42
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (calculated on anhydrous) 99.5 - 100.5 %
Appearance Conforms (see CoA/CoS)
Identification B Passes test
Appearance of solution Passes test
Readily carbonisable substances Passes test
Oxalic acid Max. 360 ppm
SO4 (Sulphate) Max. 150 ppm
Heavy metals (as Pb) Max. 10 ppm
Water Max. 1.0 %
Sulphated ash Max. 0.1 %
Residual solvents Unlikely by manuf.process


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR