Kloorivetyliuos 37%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent

Toimittaja: VWR Chemicals
Vaara

20252.290EA 27.3 EUR
20252.290 20252.244 20252.324 20252.335 20252.368 20252.448 20252.420 20252.295
Kloorivetyliuos 37%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Kloorivetyliuos
Kaava: HCl
Kiehumispiste: 110 °C (1013 hPa)
Sulamispiste: –30 °C
Tiheys: 1,18 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011324
CAS-numero: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
REACH: 01-2119484862-27

Order Now

Testitulosten erittely

Assay 35.0 - 38.0 %
Appearance Clear colourless liquid
Appearance of solution Passes test Ph.Eur.
Identification A Passes test Ph.Eur.
Identification B Passes test Ph.Eur.
Colouration Max. 10 APHA
Density (20/4) 1.174 - 1.193
Evaporation residue Max. 100 ppm
Free chlorine Max. 1 ppm
Heavy metals (as Pb) Max. 2 ppm
Ignition residue (SO4) Max. 5 ppm
Reducing substances (as O) Max. 4 ppm
Br (Bromide) Max. 5 ppm
NH4 (Ammonium) Max. 1 ppm
PO4 (Phosphate) Max. 0.5 ppm
SO3 (Sulphite) Max. 1 ppm
SO4 (Sulphate) Max. 1 ppm
Al (Aluminium) Max. 0.2 ppm
As (Arsenic) Max. 0.01 ppm
Ba (Barium) Max. 0.02 ppm
Be (Beryllium) Max. 0.01 ppm
Bi (Bismuth) Max. 0.02 ppm
Ca (Calcium) Max. 1 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.07 ppm
Cu (Copper) Max. 0.1 ppm
Fe (Iron) Max. 0.3 ppm
Ge (Germanium) Max. 0.02 ppm
Hg (Mercury) Max. 0.01 ppm
K (Potassium) Max. 0.1 ppm
Li (Lithium) Max. 0.01 ppm
Mg (Magnesium) Max. 0.1 ppm
Mn (Manganese) Max. 0.01 ppm
Mo (Molybdenum) Max. 0.02 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.04 ppm
Pb (Lead) Max. 0.02 ppm
Sr (Strontium) Max. 0.01 ppm
Ti (Titanium) Max. 0.02 ppm
Tl (Thallium) Max. 0.02 ppm
V (Vanadium) Max. 0.02 ppm
Zn (Zinc) Max. 0.5 ppm
Zr (Zirconium) Max. 0.02 ppm
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR