Xylene fast yellow 2G 60% (dye content), Sigma-Aldrich®

Toimittaja: SIGMA ALDRICH MICROSCOPY


M-Clarity™ quality level = MQ100

201987-100G
201987-100GEA 112 EUR
201987-100G
Xylene fast yellow 2G 60% (dye content), Sigma-Aldrich®
Xylene fast yellow 2G

Acid yellow-17 (C.I. 18965), also known as Disodium 2, 5-dichloro-4-(5-hydroxy-3-methyl-4-(sulphophenylazo)pyrazol-1-yl) benzenesulphonate, and Yellow 2G is an anionic, reactive textile dye. This synthetic azo dye possesses a stable aromatic structure with SO₃H and OH groups. The applications of Acid yellow-17 are seen in detergent, soap, textile, printing, and cosmetic industries.

Kaava: C₁₆H₁₂Cl₂N₄O₇S₂
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00053367
CAS-numero: 6359-98-4
EINECS: 228-819-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6dc2551f7aa18631adfaa6febfc58e0a6a410cb9&vid=6dc2551f7aa18631adfaa6febfc58e0a6a410cb9&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance (Color) Yellow
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wavelength 252- 258 nm
Extinction Coefficient ≥14,000
Extinction Coefficient c = 0.02g/L; Water ≥8,000
Carbon ≥19.5 %
Nitrogen ≥5.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR