Pinacyanol chloride, Sigma-Aldrich®

Toimittaja: SIGMA ALDRICH MICROSCOPY
Varoitus


M-Clarity™ quality level = MQ100

201715-1G 201715-250MG 201715-5G
201715-1GEA 162 EUR
201715-1G 201715-250MG 201715-5G
Pinacyanol chloride, Sigma-Aldrich®
Pinacyanol chloride

Pinacyanol chloride, (1,1'-Diethyl-2,2'-carbocyanine chloride; 2,2'-Trimethinequinocyanine chloride; Quinaldine blue).

Kaava: C₂₅H₂₅ClN₂
Sulamispiste: 270 °C (haj. lämpötila, lit.)
Storage Temperature: Room Temperature
MDL numero: MFCD00011974
CAS-numero: 2768-90-3
EINECS: 220-457-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af12451aaea8365a782670559227c19d7b76b274&vid=af12451aaea8365a782670559227c19d7b76b274&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance (Colour) Green to Very Dark Green and Green-Black
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Wavelength 559 - 565 nm
Extinction Coefficient ≥150000
Extinction Coefficient, 0.001 g/L, Ethanol ≥65000


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR