Asetoni ≥99.0% Ph. Eur.

Toimittaja: VWR Chemicals
Vaara

20165.298EA 41.7 EUR
20165.298 20165.323 20165.367 20165.460 20165.550
Asetoni ≥99.0% Ph. Eur.
Asetoni
Kaava: CH₃COCH₃
Kiehumispiste: 56,2 °C (1013 hPa)
Tiheys: 0,792 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008765
CAS-numero: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay Min. 99.0 %
Appearance Clear colourless liquid
Identification B Passes test
Identification C Passes test
Appearance of solution Passes test
Acidity or alkalinity Passes test
Relative density 0.79 - 0.793
Reducing substances Passes test
Methanol (V/V) Max. 0.05 %
Propan-2-ol (V/V) Max. 0.05 %
Benzene (V/V) Max. 2 ppm
Any other impurity (V/V) Max. 0.05 %
Matter insoluble in water Passes test
Residue on evaporation Max. 50 ppm
Water Max. 3 g/l
Residual solvents Passes test
Mesityl oxide (*) Max. 10 ppm
Statement of original manufacturer (*)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR