Etikkahappo glacial 99.8-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent

Toimittaja: VWR Chemicals
Vaara

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

20104.334EA 97.2 EUR
20104.334 20104.298 20104.243 20104.312 20104.367 20104.447 20104.551 20104.323
Etikkahappo glacial 99.8-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Etikkahappo
Kaava: H₃CCOOH
MW: 60.05 g/mol
Kiehumispiste: 118
Sulamispiste: 17
Tiheys: 1.05 g/cm³ (20 °C)
Leimahduspiste: 38.5 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00036152
CAS-numero: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 (3) II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay 99.8 - 100.5 %
Appearance Passes test Ph.Eur.
Dilution test Passes test ACS
IR Spectrum Passes test
Substances reducing dichromate Passes test ACS
Substances reducing KMnO4 Passes test
Alkalinity Max. 0.0004 meq/g
Boiling point 117 - 119 °C
Colouration Max. 10 APHA
Solidification point 16.2 - 16.6 °C
Substances coloured by H2SO4 Max. 150 APHA
Acetaldehyde Max. 2 ppm
Acetic anhydride Max. 100 ppm
Evaporation residue Max. 5 ppm
Formate Max. 0.05 %
Heavy metals (as Pb) Max. 0.5 ppm
Ignition residue (SO4) Max. 10 ppm
Water Max. 0.25 %
Cl (Chloride) Max. 0.5 ppm
NO3 (Nitrate) Max. 2 ppm
PO4 (Phosphate) Max. 0.5 ppm
SO4 (Sulphate) Max. 1 ppm
Ag (Silver) Max. 0.5 ppm
Al (Aluminium) Max. 0.03 ppm
As (Arsenic) Max. 0.01 ppm
Ba (Barium) Max. 0.01 ppm
Be (Beryllium) Max. 0.01 ppm
Bi (Bismuth) Max. 0.02 ppm
Ca (Calcium) Max. 0.2 ppm
Cd (Cadmium) Max. 0.02 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.08 ppm
Cu (Copper) Max. 0.01 ppm
Fe (Iron) Max. 0.2 ppm
Ge (Germanium) Max. 0.02 ppm
K (Potassium) Max. 0.1 ppm
Li (Lithium) Max. 0.5 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.01 ppm
Mo (Molybdenum) Max. 0.02 ppm
Na (Sodium) Max. 1 ppm
Ni (Nickel) Max. 0.1 ppm
Pb (Lead) Max. 0.02 ppm
Sr (Strontium) Max. 0.01 ppm
Ti (Titanium) Max. 0.02 ppm
Tl (Thallium) Max. 0.02 ppm
V (Vanadium) Max. 0.01 ppm
Zn (Zinc) Max. 0.05 ppm
Zr (Zirconium) Max. 0.02 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR