Etikkahappo glacial USP, Ph. Eur.

Toimittaja: VWR Chemicals
Vaara

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

20102.292EA 83.5 EUR
20102.292 20102.463 20102.320 20102.550
Etikkahappo glacial USP, Ph. Eur.
Etikkahappo

Appearance: Crystalline mass or clear, colourless, volatile liquid.


Not for use as Active Pharmaceutical Ingredient (API)


Varoitus: IMPORTANT: Material will freeze if stored below 17 °C (63°F).

Kaava: H₃CCOOH
MW: 60.05 g/mol
Kiehumispiste: 118
Sulamispiste: 17
Tiheys: 1.05 g/cm³ (20 °C)
Leimahduspiste: 38.5 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00036152
CAS-numero: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 (3) II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Conforms (see CoA/CoS)
Heavy metals (as Pb) Max. 5 ppm
Residual solvents Passes test
As (Arsenic) Max. 1 ppm
Cd (Cadmium) Max. 0.2 ppm
Co (Cobalt) Max. 0.2 ppm
Hg (Mercury) Max. 1 ppm
Ni (Nickel) Max. 0.2 ppm
Pb (Lead) Max. 0.5 ppm
V (Vanadium) Max. 0.2 ppm
Conforms to Ph.Eur./USP Passes test
Ph.Eur. specifications: .
Assay [Ph.Eur.] 99.0 --> 100.5 %
Identification A Passes test Ph.Eur.
Identification B Passes test Ph.Eur.
Appearance test Passes test Ph.Eur.
Freezing point [Ph.Eur.] Min. 14.8 °C
Reducing substances Passes test Ph.Eur.
Cl (Chloride) [Ph.Eur.] Max. 25 mg/l
SO4 (Sulphate) [Ph.Eur.] Max. 50 mg/l
Fe (Iron) [Ph.Eur.] Max. 5 ppm
Residue on evaporation [Ph.Eur.] Max. 0.01 %
USP specifications: .
Assay [USP] 99.5 --> 100.5 %
Identification Passes test USP
Limit of nonvolatile residue [USP] Max. 1.0 mg
Cl (Chloride) Passes test USP
SO4 (Sulphate) Passes test USP
Readily oxidisable substances Passes test USP
Congealing temperature [USP] Min. 15.6 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR