Drying tubes, straight
Toimittaja: Lenz Laborglas GmbH & CO.KG
05330013
201-9110EA 11.7 EUR
201-9110
Drying tubes, straight
Putket Kuivausputket
DURAN®, borosilicate glass, clear. One bulb and glass tubing connection.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes