Glass stoppers for iodine flasks, Quickfit®
Toimittaja: SciLabware
SIB24 SIB29
201-8714EA 126 EUR
201-8714 BIBBSIB29
Glass stoppers for iodine flasks, Quickfit®
Tulpat Lasi
Quickfit® iodine flask stoppers.
  • Extended handle which projects well above the liquid seal
  • Available with either 24/29 or 29/32 ground cone joints
For use with Quickfit® Iodine flasks.