Splash head, vertical, Quickfit®
Toimittaja: SciLabware
SH17/23
201-8691EA 95.5 EUR
201-8691
Splash head, vertical, Quickfit®
Tislausosat Päät
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
Usually inserted between a flask and still head to prevent 'carry-over' of raw liquid.