Adapters, cone with stopcock, Quickfit®
Toimittaja: SciLabware
MF11/2/SC MF11/3/SC MF11/4/SC MF11/1/SC
201-8596EA 104 EUR
201-8596 201-8599 201-3009 201-0263
Adapters, cone with stopcock, Quickfit®
Sovittimet Putkistoliittimet, NS-hios
Adapter, PYREX®, borosilicate glass, Quickfit® joints, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
General purpose (GP) Rotaflo® PTFE stopcock providing fine flow control and positive closure. Detachable plastic screw thread connectors allow rubber hose etc. to be fitted easily and safely.