Liebig condenser with standard ground joints
Toimittaja: SciLabware
C1/23/SC C1/12/25/SC C1/13/SC C1/14/25/SC C1/22/SC C1/44/SC C1/00 C1/11/SC C1/12/SC
201-8274EA 106 EUR
201-8274 201-8255 201-8272 201-8257 201-8273 201-8275 201-8269 201-8270 201-8271
Liebig condenser with standard ground joints
Jäähdyttimet
Condensers acc. to Liebig, PYREX® borosilicate glass, for condensing vapours. With NS socket and NS cone. Popular water-jacketed condenser suitable for distillation and reflux applications.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With detachable plastic screwthread connectors for easy and safe attachment of rubber hose. Complies with DIN 12576.