Tislauskolonni, Hempel
Toimittaja: Lenz Laborglas GmbH & CO.KG
05115201 05115303 05115304 05115503 05115504 05115301 05117304 05115804 05115803 05117201 05117301 05117504 05117804
271-3211EA 46.8 EUR
271-3211 201-0350 201-3213 LENZ05115503 271-3214 201-3212 LENZ05117304 LENZ05115804 LENZ05115803 LENZ05117201 LENZ05117301 LENZ05117504 LENZ05117804
Tislauskolonni, Hempel
Tislausosat Tislauskolonnit
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours, with cooling coil and cooling mantle. Standard ground cone and jacket. Water supplied through plastic connectors with 8 mm external Ø and GL threaded joint.
DIN 12593
Sertifikaatit: DIN 12593