Rotary evaporating flasks
Toimittaja: SciLabware
FD50/4RE FD250/4RE FD500/4RE FD1L/4RE FD2L/4RE FD250/3 FD100/3 FD100/4RE FD500/3 FD1L/3
201-8404EA 45.6 EUR
201-8404 201-8403 201-8406 201-8401 201-8009 201-8402 531-0741 201-2374 BIBBFD500/3 BIBBFD1L/3
Rotary evaporating flasks
Kolvit ja erlenmeyerit Pyöröhaihduttimen pullot
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for use with rotary evaporators. Large surface area for rapid evaporation. Robust heavy wall construction for added strength. Florentine shape allows easy removal of solid residues.