VWR® Expansion Adapters for Conical Ground Joints
Toimittaja: VWR Collection
201-1439EA 13 EUR
201-1439 201-1440 201-1445 201-1446 201-1447 201-1448
VWR® Expansion Adapters for Conical Ground Joints
Sovittimet Sovittimet, NS-hios
Adapters from small cone at the top to large socket at the bottom.
  • Made from clear borosilicate 3.3 glass
  • Excellent chemical resistance, high temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
  • Outstanding leak tightness
Sertifikaatit: Produced according to DIN 12257.