VWR®, Glass Beads
Toimittaja: VWR Collection
201-1251EA 7.2 EUR
201-1251 201-1252 201-1253 201-1254 201-1255 201-1256 201-1257 201-1258 201-1259 201-1260
VWR®, Glass Beads
Helmet ja partikkelit Lasihelmet
Soda-lime glass, solid.
  • Not suitable for heat applications
  • Available in sizes from 1 to 10 mm
Can be used for many different applications in the laboratory. In microbiology laboratories they are used for the production of single cell suspensions or for plating. In chemical laboratories they are used as filling material for distillation columns. They can also be used to clean round bottom vessels.