Gas Washing Bottle with Filter and Thread GL 45
Toimittaja: ROBU GLASFILTER
41101 41100 41102 41103 41104
201-1901EA 77.8 EUR
201-1901 201-0922 201-0923 201-0924 201-0925
Gas Washing Bottle with Filter and Thread GL 45
Pullot Kaasunpesupullot
ROBU® washing bottle with a filter disc and GL 45 thread provides efficient filtration and secure sealing, featuring a screw cap with silicone for a tight, leak-proof closure, ideal for laboratory and industrial applications.
  • Integrated VitraPOR® filter disc
  • High-quality GL 45 screw cap
  • Includes aperture for easy access to contents
  • Complete set with durable silicone washer for a secure seal
  • Made from chemical-resistant materials
  • Provides a tight and leak-proof fit
  • Reusable and easy to clean
  • Material: Borosilicate glass 3.3, plastic, silicone