Dropping funnels, conical shape, without pressure balance
Toimittaja: Lenz Laborglas GmbH & CO.KG
04453702 04457004 04454904 04455804 04473702 04474904 04475804 04457304 04452802 04472802 04477004 04477304
201-0910EA 93.7 EUR
201-0910 201-0913 201-0911 201-0912 201-0915 201-0579 201-0916 532-0051 201-0909 201-0914 532-0050 201-0917
Dropping funnels, conical shape, without pressure balance
Suppilot Tippasuppilot, NS-hios
Borosilicate glass 3.3.
  • Screw thread retaining device
  • With inscription label, without graduation
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With NS socket, NS cone and stopcock with glass or PTFE plug.