Sovittimet, syöttö, taivutetut, ilmanvaihto
Toimittaja: Lenz Laborglas GmbH & CO.KG
05268001 05268002 05268003 05268004
201-0905EA 32.7 EUR
201-0905 LENZ05268002 LENZ05268003 LENZ05268004
Sovittimet, syöttö, taivutetut, ilmanvaihto
Tislausosat Tislaussovittimet
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For highly boiling liquids, with ventilation, with NS socket and NS cone, with drain tip.