Adapters, expansion, for standard ground joints, Quickfit®
Toimittaja: SciLabware
XA41 XA21 XA32 XA42 XA54 XA43 XA31 XA52 XA53 XA10
201-0236EA 92 EUR
201-0236 201-0229 201-0235 201-0237 201-0707 201-0238 201-0230 201-0705 201-0706 201-0704
Adapters, expansion, for standard ground joints, Quickfit®
Sovittimet Sovittimet, NS-hios
Expansion adapter, PYREX® borosilicate glass, from large socket on the top to small cone at the base.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Useful for the connection of dissimilar sized ground glass joints.