Pullo, leveä suuaukko, kierrekorkilla
Toimittaja: GLASWARENFABRIK KARL HECHT
41325066 41325012 41325072 41325036 41325048 41325060 41325018 41325024 41325054 41325030
215-2825EA 136 EUR
215-2825 215-2827 201-0515 201-0516 201-0517 201-0519 201-0520 201-0522 HECH41325054 HECH41325030
Pullo, leveä suuaukko, kierrekorkilla
Pullot Lasipullot
Brown glass with black PP cap.
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
  • Round thread in accordance with DIN 168
Ideal for sampling, storing and transporting powders and pastes. Brown coloured to protect light-sensitive materials.