Tislausosat, pisaraloukku, Stutzer, bent
Toimittaja: Lenz Laborglas GmbH & CO.KG
05037022 05037033 05037044
201-0343EA 98.3 EUR
201-0343 201-0344 271-2231
Tislausosat, pisaraloukku, Stutzer, bent
Tislausosat Päät
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Two standard ground cones, inclined at an angle of 75°. Placed directly on the distillation flask to intercept droplets.