2,4-Pentaanidioni ≥99.0%, AnalaR NORMAPUR® analytical reagent

Toimittaja: VWR Chemicals
Varoitus

20092.230EA 89.4 EUR
20092.230
2,4-Pentaanidioni ≥99.0%, AnalaR NORMAPUR® analytical reagent
2,4-Pentaanidioni
Kaava: CH₃COCH₂COCH₃
Kiehumispiste: 140,4 °C (1013 hPa)
Sulamispiste: –23 °C
Tiheys: 0,96945 g/cm³ (20 °C)
Leimahduspiste: 30 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008787
CAS-numero: 123-54-6
EINECS: 204-634-0
UN: 2310
ADR: 3 (6.1),III
Merck Index: 13,00083
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c85a64bf9c7997e9ab03e421a93e7c71e57b8a3&vid=134ed3029693deef3759692be256c57b3a58b087&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (on anhydrous substance) Min. 99.0 %
Evaporation residue Max. 50 ppm
Water Max. 0.2 %
Cu (Copper) Max. 1 ppm
Fe (Iron) Max. 1 ppm
Pb (Lead) Max. 1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR