Asetoni ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent

Toimittaja: VWR Chemicals
Vaara

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

20066.310EA 32.1 EUR
20066.310 20066.365 20066.423 20066.467 20066.558 20066.296 20066.321 20066.330 20066.930
Asetoni ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Asetoni
Kaava: CH₃COCH₃
MW: 58.08 g/mol
Kiehumispiste: 56.2 °C (1013 hPa)
Sulamispiste: –95.4 °C
Tiheys: 0.792 g/cm³ (20 °C)
Leimahduspiste: <–20 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008765
CAS-numero: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Assay (on anhydrous substance) Min. 99.8 %
Appearance of solution Passes test Ph.Eur.
Aqueous solution Passes test
Identification B Passes test Ph.Eur.
Identification C Passes test Ph.Eur.
Insolubility in water Passes test
IR Spectrum Passes test
Related substances Passes test Ph.Eur.
Acidity Max. 0.0003 meq/g
Alkalinity Max. 0.0003 meq/g
Boiling point 56.0 - 56.7 °C
Colouration Max. 10 APHA
Density (20/4) 0.790 - 0.792
Density (20/20) 0.790 - 0.793
Aldehydes (as HCHO) Max. 10 ppm
Ethanol Max. 100 ppm
Evaporation residue Max. 5 ppm
Methanol Max. 0.05 %
Propan-2-ol Max. 100 ppm
Substances reducing KMnO4 (as O) Max. 2 ppm
Water Max. 0.2 %
PO4 (Phosphate) Max. 100 ppb
Al (Aluminium) Max. 0.1 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.05 ppm
Ca (Calcium) Max. 0.5 ppm
Cd (Cadmium) Max. 0.01 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.01 ppm
Fe (Iron) Max. 0.05 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.01 ppm
Na (Sodium) Max. 0.5 ppm
Ni (Nickel) Max. 0.01 ppm
Pb (Lead) Max. 0.01 ppm
Sn (Tin) Max. 0.1 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.01 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR