Asetoni ≥99.5%, GPR RECTAPUR®

Toimittaja: VWR Chemicals
Vaara

20065.327EA 47.9 EUR
20065.327 20065.293 20065.362 20065.464 20065.555 20065.470 20065.930 20065.950
Asetoni ≥99.5%, GPR RECTAPUR®
Asetoni
Kaava: CH₃COCH₃
Kiehumispiste: 56,2 °C (1013 hPa)
Sulamispiste: –95,4 °C
Tiheys: 0,792 g/cm³ (20 °C)
Leimahduspiste: <–20 °C (sulj. astia)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008765
CAS-numero: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Related substances Passes test
Boiling point 56 - 56.7 °C
Density (20/4) 0.79 - 0.793
Evaporation residue Max. 20 ppm
Assay Min. 99.5 %
Acidity Passes test
Alkalinity Passes test
Appearance of solution Passes test
Insolubility in water Passes test
IR Spectrum Passes test
Substances reducing KMnO4 (as O) Max. 2 ppm
Water Max. 0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR