Mouse IgG1 Isotype Control [clone: MOPC-21] (APC (Allophycocyanin))

Toimittaja: Tonbo Biosciences
20-4714-U100
20-4714-U100EA 91.6 EUR
20-4714-U100
Mouse IgG1 Isotype Control [clone: MOPC-21] (APC (Allophycocyanin))
Vasta-aineet
The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.

Type: Control
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: monoclonal
Clone: MOPC-21
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR