Anti-Thy1.2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 30-H12]

Toimittaja: Tonbo Biosciences
20-0903-U025 20-0903-U100
20-0903-U025EA 51.2 EUR
20-0903-U025 20-0903-U100
Anti-Thy1.2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 30-H12]
Vasta-aineet
The 30-H12 antibody reacts with mouse CD90.2 (Thy1.2). CD90.2 is a strain-specific allelic form of the GPI-linked membrane associated protein CD90 and is involved in adhesion and signal transduction. CD90.2 is expressed on thymocytes, mature T cells and neurons in mouse strains that express the CD90.2 allele (BALB/c, CBA, C3H, C57BL/6, SJL and others). 30-H12 does not react with the CD90.1 allele expressed in mouse strains such as PL and AKR.

Type: Primary
Antigen: Thy1.2 (Thy1.2)
Clonality: monoclonal
Clone: 30-H12
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR