Anti-CD3E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: OKT3]
Toimittaja: Tonbo Biosciences
20-0037-T100 20-0037-T025
20-0037-T100EA 72.5 EUR
20-0037-T100 20-0037-T025
Anti-CD3E Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: OKT3]
Vasta-aineet
The OKT3 antibody is specific for human CD3e, also known as CD3 epsilon, a 20 kDa subunit of the T cell receptor complex along with CD3 gamma and CD3 delta. These integral membrane protein chains assemble with additional chains of the T cell receptor (TCR), as well as CD3 zeta chain, to form the T cell receptor – CD3 complex. Together with co-receptors CD4 or CD8, the complex serves to recognize antigens bound to MHC molecules on antigen-presenting cells. These interactions promote T cell receptor signaling (T cell activation), inducing a number of cellular responses including proliferation,More Product Information