Dibutyltin diacetate ≥95%

Toimittaja: Thermo Fisher Scientific
Vaara

18595.18 18595.30 18595.A1 18595.14
18595.18EA 120 EUR
18595.18 18595.30 18595.A1 18595.14
Dibutyltin diacetate ≥95%
Dibutyltin diacetate

Purity: 95% CAS No.: 1067-33-0 Molecular Formula: C12H24O4Sn Formula Weight: 351.01 Flash Point: 143°(290°F) Physical Form: Liquid Density: 1.32 Refractive Index: 1.4700 Appearance: Colourless to pale yellow Odour: Acidic Melting Point: 12° Boiling Point: 142-145°/10mm Solubility: Soluble in water and most organic solvents UN No.: 2788 MDL No.: MFCD00008697


As as stabilizer in chlorinated organics, and as a catalyst for condensation reactions

Kaava: C₁₂H₂₄O₄Sn
Kiehumispiste: 142…145 °C (10 mmHg)
Sulamispiste: 12 °C
Tiheys: 1,32
Leimahduspiste: 143 °C (290 °F)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008697
CAS-numero: 1067-33-0
EINECS: 213-928-8
UN: 2788
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR