n-Butyllithium 1.6 M in hexanes, AcroSeal®

Toimittaja: Thermo Fisher Scientific
Vaara

181271000 181275000 181278000
181271000.EA 70.3 EUR
181271000. 181275000. 181278000.
n-Butyllithium 1.6 M in hexanes, AcroSeal®
n-Butyllithium
Kaava: C₄H₉Li
Sulamispiste: –95 °C
Tiheys: 0,68 g/cm³
Leimahduspiste: –18 °C
Storage Temperature: Jääkaappi
MDL numero: MFCD00009414
CAS-numero: 109-72-8
EINECS: 203-698-7
UN: 3394
ADR: 4.2,I

Order Now

Testitulosten erittely

Appearance Clear to hazy colorless to yellow Solution (May develop some turbidity or precipitate)
Titration Active base 1.49 to 1.70 M
Free base ≤1 %

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR