Vacuum pumps, modular and expandable, type N 820 G and N 840 G, LABOPORT®
Toimittaja: KNF
N840G N820G
181-0585EA 2820 EUR
181-0585 181-0584
Vacuum pumps, modular and expandable, type N 820 G and N 840 G, LABOPORT®
Pumput Vakuumipumput
The N 820 G and N 840 G basic pumps feature a control knob for manual adjustments of the flow rate, making them well suited to a wide variety of applications. This has integrated rotational speed control, the pumps are quiet, very energy efficient, and can be optimally adjusted in accordance with the application.
  • Flow rate of up to 2,04 m³/h
  • Ultimate vacuum of 6 mbar abs
  • Combination of TFM™ PTFE pump head and PTFE coated diaphragm makes these pumps ideal for working with extremely aggressive/corrosive gases and vapours
  • High level of compatibility with