Filter pump
Toimittaja: USBECK Laborgeräte
5700 5701 5710 5721 5722 5720 5751
181-0116EA 110 EUR
181-0116 181-0117 181-0118 181-0119 181-0120 USBE5720 USBE5751
Filter pump
Pumput Vesisuihkupumput
Water jet pump in nickel-plated brass.
  • Stable and solid
  • A side knob allows connection to a vacuum pipe with an interior diameter of 8 mm
  • Available with or without non-return valve
  • Supplied with water connection for pipes with an internal diameter of 14 mm or bolting for connection to a tap
  • Pressure gauge available as an option for all models