Manual, solenoid and motor valves
Toimittaja: Cytiva
18-1108-40 18-1110-89 18-1127-66 18-1108-41 18-1108-42 19-0016-01 19-0017-01 19-1994-01 19-2098-01 19-2145-01 18-1801-42
18-1108-40EA 3011 EUR
18-1108-40 18-1110-89 18-1127-66 18-1108-41 GEHE18-1108-42 19-0016-01 19-0017-01 19-1994-01 19-2098-01 19-2145-01 18-1801-42
Manual, solenoid and motor valves
Venttiilit Switching Valves
Range of manual and automated valves to meet a variety of performance requirements. They are designed for biocompatibility and years of reliable service, each may be inserted into the flow path without causing unnecessary mixing or sample dilution.
INV-907, IV-908, and PV-908 are motorised valves that are used in ÄKTAdesign platform. INV-907 is a 7-port valve (Fig 1) primarily intended for sample injection. IV-908 and PV-908 are 8-port versions for selection of columns, samples, or buffers. IV-908 can be used at pressures up to 20 bar (2 MPa, 290 psi) and is used on the low pressure sideMore Product Information