Tetrapropylammonium hydroxide 40% (w/w) vesiliuos

Toimittaja: Thermo Fisher Scientific
Vaara

17456.14 17456.22 17456.36
17456.14EA 201 EUR
17456.14 17456.22 17456.36
Tetrapropylammonium hydroxide 40% (w/w) vesiliuos
Tetrapropylammonium hydroxide

CAS No.: 4499-86-9 Molecular Formula: C12H29NO Formula Weight: 203.37 Physical Form: Liquid Density: 0.99 Refractive Index: 1.3716 Sensitivity: Air Sensitive Appearance: Colourless to pale yellow Boiling Point: 102° Beilstein Reference: 4207983 UN No.: 3267 MDL No.: MFCD00009360

Kaava: C₁₂H₂₉NO
Kiehumispiste: 102 °C
Tiheys: 0,99
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009360
CAS-numero: 4499-86-9
EINECS: 224-800-6
UN: 3267
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR