Lithium tetrachloropalladate(II)

Toimittaja: Thermo Fisher Scientific
Varoitus

17316.03 17316.06
17316.03EA 596 EUR
17316.03 17316.06
Lithium tetrachloropalladate(II)
Lithium tetrachloropalladate(II)

CAS No.: 15525-45-8 Molecular Formula: Li2PdCl4 Formula Weight: 262.09 Physical Form: Crystalline Sensitivity: Moisture Sensitive MDL No.: MFCD00011082

Kaava: Cl₄Li₂Pd
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011082
CAS-numero: 15525-45-8
EINECS: 239-567-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR