VWR®, Ultrapure Water System, A Series
Toimittaja: VWR Collection
171-1250EA 6610 EUR
171-1250 171-1249 171-1460 171-1461
VWR®, Ultrapure Water System, A Series
Vedenpuhdistusjärjestelmät Puhtaan ja ultrapuhtaan veden järjestelmät
This compact 'plug and play' lab water system is an ideal choice for users who need up to 20 litres of ultrapure Type I water or RO water (Type III) per day. Quality of the ultrapure product water meets or exceeds ASTM, CLSI, CAP, and ISO Type I water standards.
  • Simple and intuitive to dispense water via the dispenser or central console
  • Central console is an 8 inch waterproof touch screen, providing user full control and access of system performance
  • One water system can drive several dispensing units ('1 + N mode')
  • Dispensing flow rate is up to 0,5 L/min