Affinity chromatography columns, HiTrap™ Blue HP
Toimittaja: Cytiva
17-0412-01 17-0413-01
17-0412-01EA 272 EUR
17-0412-01 17-0413-01
Affinity chromatography columns, HiTrap™ Blue HP
Kromatografiakolonnit
HiTrap™ Blue HP is prepacked with blue Sepharose™ HP for capture of a wide range of biomolecules such as albumin, interferon, a broad range of nucleotide-dependent enzymes, α2-macroglobulin, coagulation factors and related proteins.
  • Can be used for depletion of albumin from serum
  • The ligand, Cibacron Blue F3G-A, is covalently attached to the highly cross-linked agarose medium and is stable over a pH range of 4 to 12 for use and 3 to 13 for cleaning
  • Can be operated with a syringe, peristaltic pump or liquid chromatography system such as ÄKTA design or FPLC