Gel filtration media, Sephadex™ G-75
Toimittaja: Cytiva
17-0050-01 17-0051-01
17-0050-01EA 438 EUR
17-0050-01 17-0051-01
Gel filtration media, Sephadex™ G-75
Proteiininpuhdistusjärjestelmät Protein Purification Resins
Sephadex™ G-75 is well established gel filtration medium for desalting and buffer exchange of large biomolecules >80 000 molecular weight.
  • Quickly desalts, removes contaminants and transfers to a new buffer in a single step.
  • Suitable for buffer exchange and cleanup of large biomolecules, >80 000 molecular weight
  • Classic gel filtration medium
Sephadex™ is a gel filtration resin prepared by crosslinking dextran with epichlorohydrin. Different types of Sephadex™ differ in their degree of cross-linking and hence in their degree of swelling and their molecular fractionation range. Sephadex™ G-75 is one of six different G-types ranging from G-10 for small molecules to G-100 for very large molecules. Sephadex™ G-75 Superfine has the smallest bead size for higher efficiency with shorter diffusion distances.