4-Bromi-1-butanoli cont. varying amounts of THF, tech.

Toimittaja: Thermo Fisher Scientific
Varoitus

16805.03 16805.06 16805.14
16805.03EA 156 EUR
16805.03 16805.06 16805.14
4-Bromi-1-butanoli cont. varying amounts of THF, tech.
4-Bromi-1-butanoli

CAS No.: 33036-62-3 Molecular Formula: C4H9BrO Formula Weight: 153.02 Storage Temperature: -30°C to -10°C Flash Point: 32°(91°F) Physical Form: Liquid Sensitivity: Moisture and Light Sensitive Appearance: Yellow Boiling Point: 63-65°/5mm UN No.: 1987 MDL No.: MFCD00015387

Kaava: BrH₂C(CH₂)₂CH₂OH
Kiehumispiste: 63…65 °C (5 mmHg)
Leimahduspiste: 32 °C (91 °F)
Storage Temperature: Pakastin
MDL numero: MFCD00015387
CAS-numero: 33036-62-3
EINECS: 251-355-5
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR