Bariumbromidi, vedetön 99%

Toimittaja: Thermo Fisher Scientific
Varoitus

16792.18 16792.30
16792.18EA 186 EUR
16792.18 16792.30
Bariumbromidi, vedetön 99%
Bariumbromidi

Purity: 99% CAS No.: 10553-31-8 Molecular Formula: BaBr2 Formula Weight: 297.14 Physical Form: Powder Density: 4.781 Appearance: White Odour: Odourless Melting Point: 850° Solubility: Soluble in water Merck Reference: 14,968 UN No.: 1564 MDL No.: MFCD00003444

Kaava: BaBr₂
Kiehumispiste: 1835 °C (1013 hPa)
Sulamispiste: 850 °C
Tiheys: 4,781
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003444
CAS-numero: 10553-31-8
EINECS: 234-140-0
UN: 1564
ADR: 6.1,III
Merck Index: 14,968

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR