Rivaroxaban Related Compounds, Reference Standard

Toimittaja: USP

1604552 1604563 1604574 1604585 1604596
1604552.EA 1580 EUR
1604552. 1604563. 1604574. 1604585. 1604596.
Rivaroxaban Related Compounds, Reference Standard
Standardiaineet ja vertailumateriaalit Organic Standards

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.


Sertifikaatit: US Pharmacopea Reference Standard

Order Now

TEKNISET TIEDOT

Kuvaus Rivaroxaban Related Compound B Rivaroxaban Related Compound D Rivaroxaban Related Compound J Rivaroxaban Related Compound H Rivaroxaban Related Compound G
Environmentally Preferable
Synonym(s) (S)-N-({2-Oxo-3-[4-(3-oxomorpholino)phenyl]oxazolidin-5-yl}methyl)acetamide 1,3-Bis({(S)-2-oxo-3-[4-(3-oxomorpholino)phenyl]oxazolidin-5-yl}methyl)urea (S)-4,5-Dichloro-N-({2-oxo-3-[4-(3-oxomorpholino)phenyl]oxazolidin-5-yl}methyl)thiophene-2-carboxamide (S)-2-({2-oxo-3-[4-(3-oxomorpholino)phenyl]oxazolidin-5-yl}methyl)isoindoline-1,3-dione
CAS-nro 1429334-00-8 1365267-35-1 1632463-24-1 1770812-37-7 446292-08-6
UN No. 3077

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR