Formaldehydi solution 35% (w/w) stabilised, meets analytical specification of BP, Ph. Eur., Riedel-de Haën™

Toimittaja: Honeywell Chemicals
Vaara

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

Stabilization: 10% Stabilised with methanol

15513-60KG 15513-2.5L 15513-1L 15513-1000KG 15513-4X2.5L
15513-60KGEA 624 EUR
15513-60KG 15513-2.5L 15513-1L 15513-1000KG 15513-4X2.5L
Formaldehydi solution 35% (w/w) stabilised, meets analytical specification of BP, Ph. Eur., Riedel-de Haën™
Formaldehydi

Beilstein Number: 1209228

Kaava: CH₂O
MW: 30.03 g/mol
Kiehumispiste: 93…96 °C (1.013 hPa)
Sulamispiste: <–15 °C
Tiheys: 1.080…1.090 g/cm³ (20 °C)
Leimahduspiste: 64 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003274
CAS-numero: 50-00-0
EINECS: 200-001-8
UN: 2209
ADR: 8 III
Merck Index: 13,04259

Order Now

Testitulosten erittely

Assay 35.0 - 38.0 %
Density (D 20/4) effective
Sulfated ash Max. 0.01 %
Acidity complying
Methanol (GC) 9.0 - 15.0 Vol.%
Residual solvents complying
Appearance of the solution complying
Autoignition Temperature 572 °F
Vapor Pressure 52 mmHg ( 37 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR